Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCHP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114159 Supplier Invitrogen™ Supplier No. PA5114159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IQEKIEQNRR AQEESLKHRE QLIRNLEEVR ELARREKEES EKLKSARKQE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Yeast: 91%.

TCHP may act as a 'capping' or 'branching' protein for keratin filaments in the cell periphery. This protein may regulate K8/K18 filament and desmosome organization mainly at the apical or peripheral regions of simple epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCHP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCHP
Gene Accession No. Q9BT92
Gene Alias A930031F18Rik; AV279199; Mitochondrial protein with oncostatic activity; mitostatin; Protein TCHP; Tchp; TpMs; trichoplein keratin filament binding; trichoplein keratin filament-binding protein; trichoplein, keratin filament binding; tumor suppressor protein
Gene Symbols TCHP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human TCHP.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84260
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.