Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TCID
SDP

Catalog No. 517950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-795-0 50 mg
51-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-795-0 Supplier Tocris Bioscience™ Supplier No. 5179/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor

TCID is a selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor (IC50 = 0.6 μM). Exhibits >100-fold selectivity for UCH-L3 over UCH-L1. Diminishes glycine transporter GlyT2 ubiquitination in brain stem and spinal cord primary neurons.

Specifications

Purity >99%
Quantity 50 mg
Product Type TCID
Molecular Weight (g/mol) 283.92
Inhibitors TCID

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.