Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCIRG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP189333
Description
TCIRG1 Polyclonal specifically detects TCIRG1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TCIRG1 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
a3, Atp6i, ATP6N1Cspecific 116-kDa vacuolar proton pump subunit, ATP6V0A3T-cell immune response cDNA 7, OC-116, OC-116 kDa, OC-116kDa, OC116Vph1, OPTB1, Osteoclastic proton pump 116 kDa subunit, Stv1, T-cell immune regulator 1, T-cell immune response cDNA7 protein, T-cell, immune regulator 1, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein aisoform 3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3, TIRC7ATPase, H+ transporting, 116kD, vacuolar proton translocating ATPase 116 kDa subunit A, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3, V-ATPase 116 kDa isoform a3, V-ATPase 116-kDa, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
10312 | |
Human, Mouse | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TCIRG1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RPADRQEENKAGLLDLPDASVNGWSSDEEKAGGLDDEEEAELVPSEVLMHQAIHTI | |
0.1 mL | |
Primary | |
Specificity of human TCIRG1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction