Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCL1A Antibody (1-21) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$192.47 - $591.65
Specifications
| Antigen | TCL1A |
|---|---|
| Clone | 43121 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018525
![]() |
Novus Biologicals
NBP14325325UG |
25 μg |
Each for $192.47
|
|
|||||
NBP14325301
![]() |
Novus Biologicals
NBP1432530.1MG |
0.1 mg |
Each for $591.65
|
|
|||||
Description
TCL1A Monoclonal specifically detects TCL1A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TCL1A | |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 8115 | |
| The immunogen for this antibody was TCL1. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 43121 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
| TCL1A | |
| IgG2b κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title