Learn More
Description
Specifications
Specifications
| Antigen | TCL1A |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | TCL1/2078 |
| Conjugate | DyLight 488 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A |
| Gene Symbols | TCL1A |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
