Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCL1A Antibody (TCL1/2078), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275750IR
Description
TCL1A Monoclonal specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TCL1A | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TCL1A | |
Recombinant human TCL1A protein fragment (around aa 2-109) (exact sequence is proprietary) (Uniprot: P56279) | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
TCL1/2078 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
Mouse | |
Protein A or G purified | |
RUO | |
T-cell leukemia/Lymphoma Protein 1A (TCL1A), also known as p14TCL1 is a product of the TCL1 gene that is involved in T-cell prolymphocytic leukemia (TPLL). T-PLL is a rare form of mature T-cell leukemia, which is consistently associated with chromosomal rearrangements characterized by the juxtaposition of the TCRA locus on chromosome 14q11 and the TCL1A gene on 14q32.13. TCL1 is overexpressed in Burkitt lymphoma, the majority of AIDS-related non-Hodgkin lymphoma-designated immunoblastic plasmacytoid lymphoma, lymphoblastic lymphoma, chronic lymphocytic leukemia, mantle cell lymphoma, follicular lymphoma, diffuse large B-cell lymphoma, and primary cutaneous B-cell lymphoma. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction