Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCL1A Antibody (TCL1/2079), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311312F
Description
TCL1A Monoclonal antibody specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TCL1A | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
TCL1/2079 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
Recombinant fragment (around aa 2-109) of human TCL1 protein (exact sequence is proprietary) | |
0.1 mL | |
Cancer | |
8115 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction