Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCL1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102316 Supplier Invitrogen™ Supplier No. PA5102316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TCL1A.

Overexpression of the TCL1 gene in humans has been implicated in the development of mature T cell leukemia, in which chromosomal rearrangements bring the TCL1 gene in close proximity to the T-cell antigen receptor (TCR)-alpha or TCR-beta regulatory elements (summarized by Virgilio et al., 1998). In normal T cells TCL1 is expressed in CD4-/CD8- cells, but not in cells at later stages of differentiation. TCL1 functions as a coactivator of the cell survival kinase AKT (Laine et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCL1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TCL1A
Gene Accession No. P56279, P56280
Gene Alias oncogene TCL1; Oncogene TCL-1; Protein p14 TCL1; T cell lymphoma breakpoint 1; T-cell leukemia/lymphoma 1A; T-cell leukemia/lymphoma protein 1A; T-cell leukemia/lymphoma protein 1A (P14 TCL1 protein) (TCL1 oncogene) (TCL-1 protein); T-cell lymphoma breakpoint 1; T-cell lymphoma-1; T-cell lymphoma-1A; TCL1; TCL-1; TCL1A
Gene Symbols Tcl1, TCL1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human TCL1A(Accession P56279), corresponding to amino acid residues R60-L110.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21432, 690575, 8115
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.