Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCL1A Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518689
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Overexpression of the TCL1 gene in humans has been implicated in the development of mature T cell leukemia, in which chromosomal rearrangements bring the TCL1 gene in close proximity to the T-cell antigen receptor (TCR)-alpha or TCR-beta regulatory elements (summarized by Virgilio et al., 1998). In normal T cells TCL1 is expressed in CD4-/CD8- cells, but not in cells at later stages of differentiation. TCL1 functions as a coactivator of the cell survival kinase AKT (Laine et al., 2000).
Specifications
| TCL1A | |
| Polyclonal | |
| Unconjugated | |
| TCL1A | |
| oncogene TCL1; Oncogene TCL-1; Protein p14 TCL1; T cell lymphoma breakpoint 1; T-cell leukemia/lymphoma 1A; T-cell leukemia/lymphoma protein 1A; T-cell leukemia/lymphoma protein 1A (P14 TCL1 protein) (TCL1 oncogene) (TCL-1 protein); T-cell lymphoma breakpoint 1; T-cell lymphoma-1; T-cell lymphoma-1A; TCL1; TCL-1; TCL1A | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8115 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P56279 | |
| TCL1A | |
| Synthetic peptide sequence (QLYPDGRYRSSD) corresponding to the internal amino acids of TCL1A (aa 77-89). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction