Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCL1A Rabbit anti-Human, Clone: TCL1/2747R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30737920UG
Description
TCL1A Monoclonal specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
TCL1A | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
Recombinant human TCL1A protein fragment (around aa 2-109) (exact sequence is proprietary) | |
20 μg | |
Cancer | |
8115 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
TCL1/2747R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction