Learn More
Invitrogen™ TCL1B Polyclonal Antibody

Description
Immunogen sequence: WQMAVHTREL LSSGQMPFSQ LPAVWQLYPG RKYRAADSSF WEIADHGQID SMEQLVLTYQ PE Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 27%.
Specifications
Specifications
| Antigen | TCL1B |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TCL1B |
| Gene Accession No. | O95988 |
| Gene Alias | Oncogene TCL1B; oncogene TCL-1B; SYN-1; Syncytiotrophoblast-specific protein; T-cell leukemia/lymphoma 1B; T-cell leukemia/lymphoma protein 1B; T-cell lymphoma/leukemia 1B; TCL1; TCL1/ MTCP1-like 1; TCL1/MTCP1-like protein 1; TCL1B; TML1 |
| Gene Symbols | TCL1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.