Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCL1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547873
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547873 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547873 Supplier Invitrogen™ Supplier No. PA547873
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human TCL1A is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

TCL1B (T Cell Leukemia/Lymphoma 1B) is a Protein Coding gene. Diseases associated with TCL1B include T-Cell Leukemia and Lymphoma. Among its related pathways are PI3K-Akt signaling pathway. An important paralog of this gene is MTCP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCL1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TCL1B
Gene Accession No. O95988
Gene Alias Oncogene TCL1B; oncogene TCL-1B; SYN-1; Syncytiotrophoblast-specific protein; T-cell leukemia/lymphoma 1B; T-cell leukemia/lymphoma protein 1B; T-cell lymphoma/leukemia 1B; TCL1; TCL1/ MTCP1-like 1; TCL1/MTCP1-like protein 1; TCL1B; TML1
Gene Symbols TCL1B
Host Species Goat
Immunogen E. coli-derived recombinant human TCL1B Ala2-Asp128 (Glu93Arg).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9623
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.