Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCL6 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524335

Catalog No. PIPA524335


Only null left
Add to Cart

Description

Description

TCL6 Polyclonal Antibody for Western Blot, Flow

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCL6
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
0
TCL6
KLH conjugated synthetic peptide between 101-130 amino acids from the C-terminal region of human TCL6.
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot
Unconjugated
TCL6
T-cell leukemia/lymphoma 6 (non-protein coding); TCL6; TNG1; TNG2
Rabbit
Antigen affinity chromatography, Protein A
RUO
27004
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.