Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TCN 201
GREENER_CHOICESDP

Catalog No. 415450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-545-0 50 mg
41-541-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-545-0 Supplier R&D Systems™ Supplier No. 4154/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. GluN1/GluN2A-selective NMDA antagonist

NMDA receptor antagonist selective for GluN1/GluN2A (formally NR1/NR2A) over GluN1/GluN2B (formally NR1/NR2B) containing receptors (pIC50 values are 6.8 and <4.3, respectively, in human recombinant GluN1/GluN2A (formally NR1/NR2A) and GluN1/GluN2B (formally NR1/NR2B) FLIPR/Ca2+ assays).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 852918-02-6
Molecular Formula C21H17ClFN3O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 461.89
Purity >99%
Molecular Weight (g/mol) 461.892
Chemical Name or Material TCN 201
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.