Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110469 Supplier Invitrogen™ Supplier No. PA5110469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EICEVSEENY IRLKPLLNTM IQSNYNRGTS AVNVVLSLKL VGIQIQTLMQ KMIQQIKYNV KSRLSDVSSG ELALIILALG VCRNAEENLI YDYHLIDKLE NKFQAEIENM EAHNGTPLTN YYQLSLDVLA LCLFNGNYST AEVVNHFTPE NKNYYFGSQF SVDTGAMAVL ALTCVKKSLI NGQIKADEGS LKNISIYTKS LVEKILSEKK ENGLIGNTFS TGEAMQALFV SSDYYNENDW NCQQTLNTVL TEISQGA.

Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TCN1
Gene Accession No. P20061
Gene Alias haptocorin; haptocorrin; HC; protein R; TC I; TC1; TC-1; TCI; TCN1; transcobalamin 1; Transcobalamin I; transcobalamin I (vitamin B12 binding protein, R binder family); transcobalamin-1
Gene Symbols TCN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-280 of human TCN1 (NP_001053.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6947
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.