Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCN2 Mouse anti-Human, Clone: 2G6D2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TCN2 |
---|---|
Clone | 2G6D2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6806
![]() ![]() |
Proteintech Group Inc
663771IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.Specifications
TCN2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TC 2, TC II, TC2, TCII, TCN2, Transcobalamin 2, Transcobalamin II | |
TCN2 | |
IgG2b | |
Protein A | |
Antibody |
2G6D2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P20062 | |
6948 | |
TCN2 Fusion Protein Ag2825 | |
Primary | |
-20°C | |
TCN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title