Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCN2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989452
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9452 Supplier Bioss Supplier No. BS9919R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the vitamin B12-binding protein family. This family of proteins, alternatively referred to as R binders, is expressed in various tissues and secretions. This plasma protein binds cobalamin and mediates the transport of cobalamin into cells. This protein and other mammalian cobalamin-binding proteins, such as transcobalamin I and gastric intrisic factor, may have evolved by duplication of a common ancestral gene. Alternative splicing results in multiple transcript variants.

Specifications

Antigen TCN2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TCN2
Gene Accession No. P20062
Gene Alias AW208754; BOS_16770; D22S676; D22S750; II; LOC100358905; macrocytic anemia; TC; TC II; TC TC D22S676; TC2; TC-2; TCII; Tcn2; Tcn-2; Tcn2p; transcobalamin 2; Transcobalamin II; transcobalamin II; macrocytic anemia; transcobalamin II-like; Transcobalamin-2; vitamin B12-binding protein 2
Gene Symbols TCN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TCN2/Transcobalamin II.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6948
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.