Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCP-1 beta Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | TCP-1 beta |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119019
|
Invitrogen™
PA5119019 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5122140
|
Invitrogen™
PA5122140 |
20 μL |
Each for $237.00
|
|
|||||
Description
TCP-1 beta Polyclonal Antibody for Western Blot, IHC (P)
TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.Specifications
TCP-1 beta | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Pig | |
CCT2 | |
99D8.1; AI528772; c-cpn; CCT; Cct1; CCT2; Ccta; CCTB; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1 subunit 2; chaperonin containing Tcp1, subunit 2 (beta); chaperonin subunit 2 (beta); chaperonin-containing T-complex polypeptide beta subunit; epididymis secretory sperm binding protein Li 100n; HEL-S-100n; hypothetical protein LOC505313; p63; PRO1633; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; Tcp-1; TCP-1-beta; Tp63; TRic | |
CCT2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P78371, P80314 | |
100157776, 10576, 12461 | |
Recombinant protein Chaperonin Containing TCP1, Subunit 2. The antigen corresponds to amino acid range 1-488 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title