Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP-1 delta Polyclonal Antibody, Invitrogen™

Catalog No. PIPA583083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583083 Supplier Thermo Scientific Supplier No. PA583083

Rabbit Polyclonal Antibody

Immunogen sequence: GSNKLVIEEA ERSIHDALCV IRCLVKKRAL IAGGGAPEIE LALRLTEYSR TLSGMESYCV RAFADAMEVI PSTLAENAGL NPISTVTELR NRHAQGEKTA GINVRKGGIS NILEELVVQP LLVSVSALTL ATETVRSILK

The chaperonin containing TCP1 (MIM 186980) complex (CCT), also called the TCP1 ring complex, consists of 2 back-to-back rings, each containing 8 unique but homologous subunits, such as CCT4. CCT assists the folding of newly translated polypeptide substrates through multiple rounds of ATP-driven release and rebinding of partially folded intermediate forms. Substrates of CCT include the cytoskeletal proteins actin (see MIM 102560) and tubulin (see MIM 191130), as well as alpha-transducin (MIM 139330) (Won et al., 1998 [PubMed 9819444]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 delta
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cct4
Gene Accession No. P50991
Gene Alias 2610204B21Rik; A45; C78323; CCT4; cct4 protein; Cctd; CCT-delta; chaperonin containing t-complex polypeptide 1, delta subunit; chaperonin containing TCP1 subunit 4; chaperonin containing Tcp1, subunit 4 (delta); chaperonin subunit 4 (delta); SRB; Stimulator of TAR RNA-binding; T complex protein 1, delta; T-complex protein 1 subunit delta; TCP-1 delta; TCP-1-delta; Unknown (protein for MGC:134500); zgc:65789; zgc:77192
Gene Symbols Cct4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCT4
Purification Method Anitgen affinity chromatography, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.