Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP-1 epsilon Monoclonal Antibody (PK/29/23/8d)
GREENER_CHOICE

Catalog No. PIMA182643
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA182643 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA182643 Supplier Invitrogen™ Supplier No. MA182643
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In Western blot, this antibody detects a band at ~60 kD in lysates of heat-shocked Hela cells. Rat anti CCT epsilon antibody, clone PK/29/23/8d recognizes the epsilon polypeptide of the CCT chaperonin molecule complex.

This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 epsilon
Applications Gel Shift, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PK/29/23/8d
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene CCT5
Gene Accession No. P48643, P80316, Q68FQ0
Gene Alias CCT5; CCTE; CCT-epsilon; chaperonin containing TCP1 subunit 5; chaperonin containing Tcp1, subunit 5 (epsilon); chaperonin subunit 5 (epsilon); epididymis secretory protein Li 69; HEL-S-69; Kiaa0098; mKIAA0098; PNAS-102; T-complex protein 1 subunit epsilon; TCP-1 epsilon; TCP-1-epsilon; TCPE; unm tf212; unm_tf212; wu:fb54h08
Gene Symbols CCT5
Host Species Rat
Immunogen Proprietary Immunogen.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100350572, 12465, 22948, 294864, 533784
Target Species Human, Mouse, Rat, Rabbit, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.