Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP-1 eta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543729 Supplier Invitrogen™ Supplier No. PA543729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RAIKNDSVVA GGGAIEMELS KYLRDYSRTI PGKQQLLIGA YAKALEIIPR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 100%.

The TCP1 eta gene encodes a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 5 and 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 eta
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCT7
Gene Accession No. Q99832
Gene Alias AA408524; AL022769; CCT7; CCTETA; CCT-eta; CCTH; Cctz; chaperonin containing t-complex polypeptide 1, eta subunit; chaperonin containing t-complex subunit eta; chaperonin containing TCP1 subunit 7; chaperonin containing Tcp1, subunit 7 (eta); chaperonin subunit 7 (eta); HIV-1 Nef interacting protein; HIV-1 Nef-interacting protein; MGC110985; NIP7-1; T-complex protein 1 subunit eta; T-complex protein 1 subunit eta, N-terminally processed; TCP1ETA; TCP-1-eta
Gene Symbols CCT7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CCT7.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10574
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.