Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP-1 theta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553830 Supplier Invitrogen™ Supplier No. PA553830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPNLVCCSAK NLRDIDEVSS LLRTSIMSKQ YGNEVFLAKL IAQACVSIFP DSGHFNVDNI RVCKILGSGI SSSSVLHGMV FKKETEGDVT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

TCP-1 theta is theta subunit of the CCT (chaperonin containing TCP1 complex) chaperonin molecule which is abundant in the eukaryotic cytosol and may be involved in the transport and assembly of the newly synthesized proteins. Molecular chaperones assist in the protein folding upon ATP hydrolysis and TCP-1 theta may play a role in the assembly of the BBsome, a complex that is involved in ciliogenesis regulating transport vesicles to the cilia. Further, TCP-1 theta is also involved in the folding of actin and tubulin. The intact CCT complex is composed of eight polypeptides in a double ring structure. CCT is important within cells in the folding of the VHL tumor suppressor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 theta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCT8
Gene Accession No. P42932, P50990
Gene Alias AI132397; bdav; bette davis; C21orf112; Cct8; Cctq; CCT-theta; Chaperonin containing T-complex polypeptide 1 subunit 8; chaperonin containing TCP1 subunit 8; chaperonin containing TCP1, subunit 8 (theta); chaperonin subunit 8 (theta); D21S246; fa22h09; KIAA0002; PRED71; renal carcinoma antigen NY-REN-15; T-complex protein 1 subunit theta; TCP-1-theta; Tcpq; wu:fa22h09; zgc:56059
Gene Symbols CCT8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCP-1 theta. Recombinant protein control fragment (Product #RP-91817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10694, 12469, 288305
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.