Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP-1 zeta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562104 Supplier Invitrogen™ Supplier No. PA562104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVTGDGTTSN VLIIGELLKQ ADLYISEGLH PRIIAEGFEA AK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.

The protein TCP-1 (t complex polypeptide 1) is a subunit of the heterooligomeric complex CCT (chaperonin containing TCP-1) present in the eukaryotic cytosol. The CCT of eukaryotic cytosol is composed of eight different subunit species, TCP-1 alpha, beta, gamma, delta, epsilon, zeta, eta and theta, each encoded by a different gene. Two zeta subunits have been described: TCP-1 zeta (also designated TCP-1 zeta1) and TCP-1 zeta2. TCP-1 subunits are proposed to have independent functions in folding its in vivo substrates, the actins and tubulins. TCP-1 was first identified in the mouse as relevant for tail-less and embryonic lethal phenotypes. Sequences homologous to TCP-1 have been isolated in several other species, and the yeast TCP-1 has been shown to encode a molecular chaperone for Actin and Tubulin. TCP-1 found in mammalian cells and yeast plays an important role in the folding of cytosolic proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 zeta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCT6A
Gene Accession No. P40227
Gene Alias acute morphine dependence related protein 2; acute morphine dependence-related protein 2; amino acid transport defect-complementing; CCT zeta1; CCT- zeta1; CCT- zeta-1; CCT zeta-1; Cct6; Cct6a; Cctz; Cctz1; Cctz-1; CCT-zeta; CCT-zeta-1; chaperonin containing T-complex subunit 6; chaperonin containing TCP-1; chaperonin containing TCP1 subunit 6A; chaperonin containing Tcp1, subunit 6A (zeta 1); chaperonin containing Tcp1, subunit 6a (zeta); chaperonin subunit 6a (zeta); histidine transp; histidine transport regulator 3; HTR3; MoDP-2; T-complex protein 1 subunit zeta; T-complex protein 1, zeta subunit; TCP 1 zeta; TCP1 zeta; TCP-1- zeta; TCP1- zeta; TCP-1-zeta; Tcp20; TCPZ; TTCP20
Gene Symbols CCT6A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCP-1 zeta-2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 908
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.