Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCT6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568388
Description
CCT6B Polyclonal Antibody for Western Blot, IHC (P)
Molecular chaperone; assists the folding of proteins upon ATP hydrolysis. Known to play a role, in vitro, in the folding of actin and tubulin.
Specifications
CCT6B | |
Polyclonal | |
Unconjugated | |
Cct6b | |
Cct6b; Cctz2; Cctz-2; CCT-zeta-2; CCTzeta-2; CCT-zeta-like; chaperonin containing TCP1 subunit 6B; chaperonin containing TCP1, subunit 6B (zeta 2); chaperonin containing Tcp1, subunit 6b (zeta); chaperonin subunit 6b (zeta); chaperonin-containing T-complex polypeptide 1, subunit 6B; T-complex protein 1 subunit zeta-2; TCP-1-zeta-2; TCP-1-zeta-like; Testis-specific protein TSA303; testis-specific Tcp20; TSA303 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10693 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q92526 | |
Cct6b | |
Synthetic protein corresponding to residues near the C terminal of human TCP-1 zeta-2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction