Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100432 Supplier Thermo Scientific Supplier No. PA5100432
Add to Cart
Add to Cart

CCT6B Polyclonal Antibody

CCT6B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCT6B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cct6b
Gene Accession No. Q61390, Q92526
Gene Alias Cct6b; Cctz2; Cctz-2; CCT-zeta-2; CCTzeta-2; CCT-zeta-like; chaperonin containing TCP1 subunit 6B; chaperonin containing TCP1, subunit 6B (zeta 2); chaperonin containing Tcp1, subunit 6b (zeta); chaperonin subunit 6b (zeta); chaperonin-containing T-complex polypeptide 1, subunit 6B; T-complex protein 1 subunit zeta-2; TCP-1-zeta-2; TCP-1-zeta-like; Testis-specific protein TSA303; testis-specific Tcp20; TSA303
Gene Symbols Cct6b
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCT6B(Accession Q92526), corresponding to amino acid residues I275-S325.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10693, 12467, 363658
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.