Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP1 alpha Antibody (23c (CTA-123)), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250328B
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TCP1 alpha | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
TCP1 | |
C-terminal half of the full length murine TCP protein | |
0.1 mL | |
Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
6950 | |
Mouse, Rat, Canine, Chicken, Hamster | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
23c (CTA-123) | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Does not recognize primate, xenopus or yeast TCP-1. | |
Store at 4C in the dark. | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction