Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP1 alpha Antibody (23c (CTA-123)) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2503280.025ML
Description
TCP1 alpha Monoclonal antibody specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
TCP1 alpha | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
C-terminal half of the full length murine TCP protein | |
0.025 mL | |
Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
6950 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2c |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
23c (CTA-123) | |
Unconjugated | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat, Canine, Chicken, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction