Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP1 delta (CCT4) Monoclonal Antibody (OTI1A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF810896


Description
TCP1 delta (CCT4) Monoclonal antibody specifically detects TCP1 delta (CCT4) in Human, Mouse samples. It is validated for Western BlotSpecifications
TCP1 delta (CCT4) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O35737, P31943 | |
Mouse | |
Protein A/G | |
RUO | |
3187, 59013 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI1A3 | |
Unconjugated | |
Hnrnph1 | |
2610204B21Rik; A45; C78323; CCT4; cct4 protein; Cctd; CCT-delta; chaperonin containing t-complex polypeptide 1, delta subunit; chaperonin containing TCP1 subunit 4; chaperonin containing Tcp1, subunit 4 (delta); chaperonin subunit 4 (delta); SRB; Stimulator of TAR RNA-binding; T complex protein 1, delta; T-complex protein 1 subunit delta; TCP-1 delta; TCP-1-delta; Unknown (protein for MGC:134500); zgc:65789; zgc:77192 | |
Full length human recombinant protein of human HNRNPH1 (NP_005511) produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction