Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP1 eta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578956
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578956 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578956 Supplier Invitrogen™ Supplier No. PA578956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human CACO-2 whole cell, human 293T whole cell, human HepG2 whole cell, human MCF-7 whole cellhuman HEL whole cell, human U251 whole cell, human K562 whole cell, rat C6 whole cell, mouse Neuro-2a whole cell. ICC/IF: U20S cell. Flow: HepG2 cell.

The TCP1 eta gene encodes a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 5 and 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP1 eta
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene CCT7
Gene Accession No. P80313, Q99832
Gene Alias AA408524; AL022769; CCT7; CCTETA; CCT-eta; CCTH; Cctz; chaperonin containing t-complex polypeptide 1, eta subunit; chaperonin containing t-complex subunit eta; chaperonin containing TCP1 subunit 7; chaperonin containing Tcp1, subunit 7 (eta); chaperonin subunit 7 (eta); HIV-1 Nef interacting protein; HIV-1 Nef-interacting protein; MGC110985; NIP7-1; T-complex protein 1 subunit eta; T-complex protein 1 subunit eta, N-terminally processed; TCP1ETA; TCP-1-eta
Gene Symbols CCT7
Host Species Rabbit
Immunogen E. coli-derived human CCT7 recombinant protein (Position: Q30-D307).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10574, 12468, 297406
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.