Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP1 Monoclonal Antibody (91a), PerCP
GREENER_CHOICE

Catalog No. PIMA545720
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545720 Supplier Invitrogen™ Supplier No. MA545720
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

1 μg/mL of MA5-45720 was sufficient for detection of TCP1 alpha in 20 μg of 3T3 cell lysate by colorimetric immunoblot analysis using Goat anti-rat IgG:HRP as the secondary antibody. Detects approximately 60kDa. Also detects approximately 92kDa. Cross reactivity with human HSP60 has been observed with this antibody in immunoblot analysis. Reacts weakly with Saccharomyces cerevisiae, consistent with the epitope sequence being AKLRS (instead of AKLRA). In C. elegans, it reacts with TCP1 alpha and another CCT subunit. In plants, it recognizes TCP1 of Pisum sativum, and the sequence of Arabidopsis thalania TCP1 over the region of the epitope AKLRA. It has also been shown that it reacts with a subunit of a specialized chaperonin which folds phytochrome.

The T Complex Polypeptide 1 (TCP-1) is approximately 60 kDa protein constitutively expressed in almost all eukaryotic cells, and is upregulated during spermatogenesis. It is found in the cytosol as a subunit of a hetero-oligomeric chaperone that is known to be involved in the folding of actin and tubulin. The family of proteins termed chaperonins act to recognize and stabilize polypeptide intermediates during folding, assembly and disassembly, and share many characteristics with Heat Shock Protein 70 (HSP70) including high abundance, induction by environmental stress, and ATPase activity. The chaperonin family includes the mitochondrial HSP60, Escherichia coli GroEL, the plastid Rubisco-subunit binding protein, and the archaebacterial protein TF55. The TCP-1 sequence shows nearly 40% identity to TF55, but only minimal similarity to HSP60 and GroEL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP1
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 91a
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene TCP1
Gene Accession No. P11983, P12612, P17987, P18279, P28480, Q32L40
Gene Alias 65 kDa antigen; AI528772; c-cpn; CCT; Cct1; Ccta; CCTalpha; CCT-alpha; chaperonin-containing T-complex alpha subunit TCP1; chaperonin-containing T-complex polypeptide alpha subunit; cytosolic chaperonin containing t-complex polypeptide 1; D6S230E; hypothetical protein; p63; tailless complex polypeptide 1; tailless complex polypeptide 1A; Tailless complex polypeptide 1B; t-complex 1; t-complex polypeptide 1; t-complex protein 1; T-complex protein 1 alpha subunit-like protein; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; T-complex protein 1, alpha subunit; Tcp1; Tcp-1; TCP-1-A; TCP-1-alpha; TCP-1-B; Tp63; TRic; Unknown (protein for MGC:133746); YD8142.13; YD8142B.04; YDR212W
Gene Symbols TCP1
Host Species Rat
Immunogen Recombinant Mouse TCP1 alpha protein fragment (carboxy terminal region).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100153485, 100302435, 100689476, 100713821, 21454, 24818, 484064, 512043, 6950, 851798
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Drosophila, Yeast, Yeast, C. elegans
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.