Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tcp1 Rat anti-Human, Mouse, Biotin, Clone:23c, Abnova™

Catalog No. 89333360 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-360 Supplier Abnova Corporation Supplier No. MAB16901

Rat monoclonal antibody raised against partial recombinant mouse Tcp1.

Specifications

Antigen Tcp1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23c
Conjugate Biotin
Description t-complex protein 1
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunoprecipitation
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Tcp1
Gene Alias AI528772, CCT, Cct1, Ccta, TRic, Tcp-1, Tp63, c-cpn, p63
Host Species Rat
Immunogen Purified recombinant mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp 1b gene. Detects the COOH group.
Quantity 100 μg
Gene ID (Entrez) 21454
Target Species Human, Mouse
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.