Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tcp1, Rat anti-Mouse, ATTO488, Clone: 23c, Abnova™

Catalog No. 89338641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-641 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-641 Supplier Abnova Corporation Supplier No. MAB18716

Rat Monoclonal Antibody

Specifications

Antigen Tcp1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23c
Conjugate Atto 488
Dilution Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene t-complex protein 1
Gene Alias AI528772/CCT/Cct1/Ccta/TRic/Tcp-1/Tp63/c-cpn/p63
Gene Symbols Tcp1
Host Species Rat
Immunogen Purified recombinant mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp 1b gene expressed in E. coli. Detects the COOH group.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21454
Test Specificity It detects ∼60kDa. Also detects ∼92kDa. The addition of an alanine (LDDA COOH) prevents binding in ELISA assays to immobilized synthetic peptide sequences.
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.