Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP10 Antibody (OTI2C2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274474R
Description
TCP10 Monoclonal specifically detects TCP10 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| TCP10 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TCP10 | |
| Full length recombinant protein of human TCP10 (NP_004601) produced in 293T cell. | |
| 0.1 mL | |
| 6953.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2C2 | |
| Western Blot, Immunohistochemistry | |
| MGC34049, t-complex 10 (a murine tcp homolog), t-complex 10 (mouse), t-complex 10 homolog (mouse), T-complex protein 10A homolog, TCP10A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction