Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCP10 Monoclonal Antibody (OTI2C2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527090
Description
TCP10 Monoclonal Antibody for Western Blot, IHC (P)
The exact function of TCP10 remains unknown.
Specifications
TCP10 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
0 | |
Tcp10 | |
Full length human recombinant protein of TCP10 produced in 293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C2 | |
Unconjugated | |
Tcp10 | |
t-complex 10; t-complex 10 (a murine tcp homolog); t-complex 10 homolog; T-complex protein 10A homolog; TCP10; TCP10A | |
Mouse | |
Affinity Chromatography | |
RUO | |
6953 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction