Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCP10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542796
Description
Peptide sequence: MLEGQLEAGE PKEGTHPEDP CPGAGAAMEK TPAAAEVPRE DSNAGEMPSL Sequence homology: Human: 100%.
The exact function of TCP10 remains unknown.
Specifications
TCP10 | |
Polyclonal | |
Unconjugated | |
Tcp10 | |
t-complex 10; t-complex 10 (a murine tcp homolog); t-complex 10 homolog; T-complex protein 10A homolog; TCP10; TCP10A | |
Rabbit | |
Affinity chromatography | |
RUO | |
6953 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
Tcp10 | |
Synthetic peptide directed towards the N-terminal of human TCP10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction