Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP10L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542797 Supplier Invitrogen™ Supplier No. PA542797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ASPHAGQESH TLALEPAFGK ISPLSADEET IPKYAGHKNQ SATLLGQRSS Sequence homology: Human: 100%.

TCP10L is a human leucine zipper protein with transcription inhibition activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP10L
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCP10L
Gene Accession No. Q8TDR4
Gene Alias C21orf77; PRED77; t-complex 10 (a murine tcp homolog)-like; T-complex 10A-2; t-complex 10-like; T-complex protein 10A homolog 1; T-complex protein 10A homolog 2; T-complex protein 10A-1; T-complex protein 10A-2; TCP10A-2; TCP10L; TCP10-like
Gene Symbols TCP10L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TCP10L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140290
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.