Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP10L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583734 Supplier Invitrogen™ Supplier No. PA583734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQLEARDPKE GTHPEDPCPG AGAVMEKT Highest antigen sequence indentity to the following orthologs: Mouse - 43%, Rat - 38%.

TCP10L is a human leucine zipper protein with transcription inhibition activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP10L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCP10L
Gene Accession No. Q8TDR4
Gene Alias C21orf77; PRED77; t-complex 10 (a murine tcp homolog)-like; T-complex 10A-2; t-complex 10-like; T-complex protein 10A homolog 1; T-complex protein 10A homolog 2; T-complex protein 10A-1; T-complex protein 10A-2; TCP10A-2; TCP10L; TCP10-like
Gene Symbols TCP10L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCP10L. Recombinant protein control fragment (Product #RP-99587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140290
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.