Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561708 Supplier Invitrogen™ Supplier No. PA561708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LILIEKELAE LGWKFLNLMH HNQQVFGPYY AEILKH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.

Plays a role in the process of sperm capacitation and acrosome reactions. Probable receptor for the putative fertilization-promoting peptide (FPP) at the sperm membrane that may modulate the activity of the adenylyl cyclase cAMP pathway. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCP11
Gene Accession No. Q8WWU5
Gene Alias D17Ken1; D6S230E; fertilization-promoting peptide receptor; FPPR; t-complex 11; t-complex 11 (a murine tcp homolog); t-complex 11 homolog; t-complex 11, testis-specific; t-complex protein 11; T-complex protein 11 homolog; Tcp11; Tcp-11; testis secretory sperm-binding protein Li 222n; testis-specific protein PBS13
Gene Symbols TCP11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCP11. Recombinant protein control fragment (Product #RP-99287).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.