Learn More
Invitrogen™ TCP11L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110470
Description
Immunogen sequence: MPFNGEKQCV GEDQPSDSDS SRFSESMASL SDYECSRQSF ASDSSSKSSS PASTSPPRVV TFDEVMATAR NLSNLTLAHE IAVNENFQLK QEALPEKSLA GRVKHIVHQA FWDVLDSELN ADPPEFEHAI KLFEEIREIL LSFLTPGGNR LRNQICEVLD TDLIRQQAEH SAVDIQGLAN YVISTMGKLC APVRDNDIRE LKATGNIVEV LRQIFHVLDL MQMDMANFTI MSLRPHLQRQ LVEYERTKFQ EILEETPSAL DQTTEWIKES VNEELFSLSE SALTPGAENT SKPSLSPTLV LNNSYLKLLQ WDYQKKELPE TLMTDGARLQ ELTEKLNQLK.
TCP11L2 is a protein coding gene.
Specifications
TCP11L2 | |
Polyclonal | |
Unconjugated | |
TCP11L2 | |
t-complex 11 like 2; t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2; TCP11L2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
255394 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.28 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8N4U5 | |
TCP11L2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human TCP11L2 (NP_689985.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.