Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR alpha Antibody (34), Alexa Fluor™ 350 - Chimeric, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP252684AF350

Catalog No. NP22684A30


Only null left
Add to Cart

Description

Description

TCR alpha Monoclonal specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

TCR alpha
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
TRA
Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11
Primary
This antibody binds to the T-cell receptor alpha chains.
Store at 4°C in the dark.
Western Blot, Flow Cytometry, ELISA
34
Western Blot, Flow Cytometry, ELISA
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Rabbit
0.1 mL
6955
Human
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.