Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha Antibody (34) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$269.00 - $584.50
Specifications
Antigen | TCR alpha |
---|---|
Clone | 34 |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25268425
![]() |
Novus Biologicals
NBP2526840.025MG |
0.025 mg |
Each for $269.00
|
|
|||||
NBP25268402
![]() |
Novus Biologicals
NBP2526840.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
TCR alpha Monoclonal specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
TCR alpha | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
TRA | |
IgG | |
Protein A or G purified | |
This antibody binds to the T-cell receptor alpha chains. |
34 | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
6955 | |
Purified recombinant soluble 85.33 scTCR. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title