Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha Antibody (F1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222493
Description
TCR alpha Monoclonal specifically detects TCR alpha in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
TCR alpha | |
Monoclonal | |
0.15 mg/mL | |
Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:50, Immunohistochemistry-Frozen 1:10 - 1:2000 | |
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This antibody reacts with Human and Primate. | |
Store at 4C. Do not freeze. | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
F1 | |
Unconjugated | |
P01848 | |
TRA | |
Human TCR V alpha-2 | |
200 μg | |
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
6955 | |
Human, Primate | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction