Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha/beta Antibody (R73), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212941AF488
Description
TCR alpha/beta Monoclonal specifically detects TCR alpha/beta in Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TCR alpha/beta | |
Monoclonal | |
Alexa Fluor 488 | |
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal R73 recognizes TCR alpha/beta, the dominant subtype of T cell receptor expressed in peripheral blood. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
R73 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
TRA | |
Rat T blasts and erythrocytes | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
6955 | |
Rat, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction