Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha/beta Antibody (R73) - Low Endotoxin and Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305296
This item is not returnable.
View return policy
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Rat, Non-human Primate samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, ImmunocytochemistrySpecifications
TCR alpha/beta | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry 1-4 ug/mL, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunocytochemistry | |
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
Rat T blasts and erythrocytes | |
0.1 mg | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
6955 | |
Store at 4°C. Do not freeze. |
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunocytochemistry | |
R73 | |
Unconjugated | |
Low endotoxin azide free phosphate buffered saline (PBS) solution, 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test. | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Rat, Non-human Primate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction