Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha/beta Antibody (R73), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212941PECY55
Description
TCR alpha/beta Monoclonal specifically detects TCR alpha/beta in Rat, Primate samples. It is validated for Flow Cytometry.Specifications
TCR alpha/beta | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
TRA | |
Rat T blasts and erythrocytes | |
0.1 mL | |
Primary | |
The mouse monoclonal R73 recognizes TCR alpha/beta, the dominant subtype of T cell receptor expressed in peripheral blood. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
R73 | |
Flow Cytometry | |
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
6955.0 | |
Rat, Primate | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction