Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha/beta Monoclonal Antibody (IP26), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | TCR alpha/beta |
---|---|
Clone | IP26 |
Concentration | 0.8 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H086T03B01A
![]() |
LIFE TECHNOLOGIES
H086T03B01A |
100 Tests |
Each for $430.00
|
|
|||||
H086T02B01A
![]() |
LIFE TECHNOLOGIES
H086T02B01A |
25 Tests |
Each for $194.00
|
|
|||||
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Flow CytometrySpecifications
TCR alpha/beta | |
0.8 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P01848, P04435, P0DSE1, P0DSE2 | |
28589, 28755, 6955, 6957 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
IP26 | |
Flow Cytometry | |
NovaFluor Blue 510 | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA | |
IgG1 κ | |
Affinity Chromatography | |
TRAC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title