Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR beta Antibody (8A3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222486
Description
TCR beta Monoclonal specifically detects TCR beta in Human, Mouse, Primate, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
TCR beta | |
Monoclonal | |
0.15 mg/mL | |
Western Blot 1:100 - 1:2000, Flow Cytometry 2 ug, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
Tcrb | |
Human TCR beta chain constant region | |
200 μg | |
Primary | |
This antibody reacts with Human. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
8A3 | |
Unconjugated | |
T cell receptor beta chain, TCRB, TCRbeta, Tib | |
Mouse | |
Protein G purified | |
RUO | |
21577 | |
Human, Mouse, Primate, Yeast | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction