Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR beta Antibody (H57-597), DyLight 350 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320118UV
Description
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
TCR beta | |
Monoclonal | |
DyLight 350 | |
T cell receptor beta chain, TCRB, TCRbeta, Tib | |
Hamsters were immunised with affinity purified DO-11.10 TCR, and produced detectable anti-TCR antibodies which were judged by the ability of the anti-sera to immunoprecipitate TCR from surface labelled DO-11.10 T cell hybridomas. Hamster 57 was selected for hybridisation, and B cell hybridomas were produced using polyethylene glycol fusion of hamster splenic B cells to the mouse myeloma variant. Selected mAb were screened after subsequent culturing for their ability to immunoprecipitate TCR from detergent lysates of surface-radiolabelled murine T cell hybridomas. | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
21577 | |
Store at 4°C in the dark. | |
IgG λ |
Flow Cytometry, Immunohistochemistry | |
H57-597 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction