Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR beta chain Armenian Hamster anti-Mouse, violetFluor™ 450, Clone: H57-597, MilliporeSigma™

Catalog No. MABF1459 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1459 100 μg
MABF1458 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1459 Supplier MilliporeSigma Supplier No. MABF1459
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

This armenian hamster monoclonal Anti-TCR beta chain Antibody (mouse), violetFluor™ 450, clone H57-597 for the detection of TCR beta chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR beta chain
Applications Flow Cytometry
Classification Monoclonal
Clone H57-597
Concentration Please refer to lot specific datasheet.
Conjugate Violet Fluor 450 (V450)
Gene Symbols Immunoglobulins & Immunology
Host Species Armenian Hamster
Immunogen Immunogen information for this antibody is proprietary information.
Purification Method Purified by size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.