Learn More
Invitrogen™ TCR beta Monoclonal Antibody (H57-597)

Description
MA1-70045 detects alpha/beta TCR in mouse samples. MA1-70045 has been successfully used in flow cytometry, immunoprecipitation, immunohistochemistry (frozen sections), and Western blot procedures. The MA1-70045 immunogen is affinity purified D0 -11.10 TCR. Stable at 4°C. For long term storage, aliquot and freeze unused portions at -20°C in volumes appropriate for single usage. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Antigen | TCR beta |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | H57-597 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Tcrb |
| Gene Accession No. | 0 |
| Gene Alias | bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; T cell receptor beta chain; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; Tcrb; TCRbeta; TCRD; Tib; TRA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.